Assay Details
| Gene Symbol: PXMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710800.3 | XP_002710846.1 | 3 - 4 | 446 | 67 | |
| XM_017341675.1 | XP_017197164.1 | 4 - 5 | 600 | 67 | ||
More Information
Set Membership: |
|

PXMP4
Genomic Map