Assay Details
| Gene Symbol: DOP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263155.2 | XP_008261377.2 | 20 - 21 | 5258 | 137 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DOP1A| Gene Symbol: DOP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263155.2 | XP_008261377.2 | 20 - 21 | 5258 | 137 | |
Set Membership: |
|