Assay Details
| Gene Symbol: VAMP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722201.3 | XP_002722247.1 | 2 | 445 | 73 | |
| XM_017338149.1 | XP_017193638.1 | 2 | 445 | 73 | ||
More Information
Set Membership: |
|

VAMP7
Genomic Map