Assay Details
| Gene Symbol: LOC100343730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017345680.1 | XP_017201169.1 | 5 - 6 | 679 | 82 | |
| XM_017345681.1 | XP_017201170.1 | 5 - 6 | 679 | 82 | ||
| XM_017345682.1 | XP_017201171.1 | 5 - 6 | 667 | 82 | ||
| XM_017345683.1 | XP_017201172.1 | 5 - 6 | 679 | 82 | ||
| XM_017345684.1 | XP_017201173.1 | 5 - 6 | 667 | 82 | ||
More Information
Set Membership: |
|

LOC100343730
Genomic Map