Assay Details
| Gene Symbol: LOC100343730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017345680.1 | XP_017201169.1 | 3 - 4 | 478 | 92 | |
| XM_017345681.1 | XP_017201170.1 | 3 - 4 | 478 | 92 | ||
| XM_017345683.1 | XP_017201172.1 | 3 - 4 | 478 | 92 | ||
More Information
Set Membership: |
|

LOC100343730
Genomic Map