Assay Details
Gene Symbol: LOC100343730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017345680.1 | XP_017201169.1 | 4 - 5 | 610 | 106 | |
XM_017345681.1 | XP_017201170.1 | 4 - 5 | 610 | 106 | ||
XM_017345682.1 | XP_017201171.1 | 4 - 5 | 598 | 106 | ||
XM_017345683.1 | XP_017201172.1 | 4 - 5 | 610 | 106 | ||
XM_017345684.1 | XP_017201173.1 | 4 - 5 | 598 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
