Assay Details
| Gene Symbol: PMS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017339082.1 | XP_017194571.1 | 773 | 81 | ||
| XM_017339083.1 | XP_017194572.1 | 841 | 81 | |||
| XM_017339084.1 | XP_017194573.1 | 331 | 81 | |||
More Information
Set Membership: |
|

PMS2
Genomic Map