Assay Details
| Gene Symbol: POLR1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262957.2 | XP_008261179.1 | 6 - 7 | 719 | 57 | |
| XM_017345151.1 | XP_017200640.1 | 6 - 7 | 692 | 57 | ||
More Information
Set Membership: |
|

POLR1C
Genomic Map