Assay Details
| Gene Symbol: DENND2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708797.3 | XP_002708843.1 | 5 - 6 | 1883 | 58 | |
| XM_008265895.2 | XP_008264117.1 | 6 - 7 | 2029 | 58 | ||
| XM_017345235.1 | XP_017200724.1 | 5 - 6 | 1698 | 58 | ||
| XM_017345245.1 | XP_017200734.1 | 4 - 5 | 410 | 58 | ||
| XM_017345247.1 | XP_017200736.1 | 3 - 4 | 1176 | 58 | ||
More Information
Set Membership: |
|

DENND2B
Genomic Map