Assay Details
| Gene Symbol: DENND2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708797.3 | XP_002708843.1 | 7 - 8 | 2197 | 74 | |
| XM_008265895.2 | XP_008264117.1 | 8 - 9 | 2343 | 74 | ||
| XM_017345235.1 | XP_017200724.1 | 7 - 8 | 2012 | 74 | ||
| XM_017345245.1 | XP_017200734.1 | 6 - 7 | 724 | 74 | ||
| XM_017345247.1 | XP_017200736.1 | 5 - 6 | 1490 | 74 | ||
More Information
Set Membership: |
|

DENND2B
Genomic Map