Assay Details
| Gene Symbol: CTSO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716884.3 | XP_002716930.1 | 7 - 8 | 952 | 74 | |
| XM_008267285.2 | XP_008265507.1 | 7 - 8 | 878 | 74 | ||
| XM_008267286.2 | XP_008265508.1 | 6 - 7 | 785 | 74 | ||
More Information
Set Membership: |
|

CTSO
Genomic Map