Assay Details
| Gene Symbol: DHTKD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717511.3 | XP_002717557.1 | 2 - 3 | 442 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DHTKD1| Gene Symbol: DHTKD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717511.3 | XP_002717557.1 | 2 - 3 | 442 | 55 | |
Set Membership: |
|