Assay Details
| Gene Symbol: ACTR3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263927.2 | XP_008262149.1 | 11 - 12 | 1259 | 59 | |
| XM_008263929.2 | XP_008262151.1 | 10 - 11 | 1138 | 59 | ||
| XM_008263930.2 | XP_008262152.1 | 10 - 11 | 1041 | 59 | ||
More Information
Set Membership: |
|

ACTR3B
Genomic Map