Assay Details
| Gene Symbol: TXNL4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711686.3 | XP_002711732.1 | 2 - 3 | 362 | 189 | |
| XM_008257555.2 | XP_008255777.1 | 2 - 3 | 373 | 189 | ||
| XM_008257556.2 | XP_008255778.1 | 2 - 3 | 506 | 189 | ||
More Information
Set Membership: |
|

TXNL4B
Genomic Map