Assay Details
| Gene Symbol: FAXDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710178.3 | XP_002710224.2 | 8 - 9 | 986 | 62 | |
| XM_008255297.2 | XP_008253519.1 | 9 - 10 | 1197 | 62 | ||
More Information
Set Membership: |
|

FAXDC2
Genomic Map