Assay Details
| Gene Symbol: LOC100356693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710484.3 | XP_002710530.3 | 3 - 4 | 475 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100356693| Gene Symbol: LOC100356693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710484.3 | XP_002710530.3 | 3 - 4 | 475 | 115 | |
Set Membership: |
|