Assay Details
| Gene Symbol: PPP2R5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717413.3 | XP_002717459.1 | 10 - 11 | 1750 | 74 | |
| XM_017347800.1 | XP_017203289.1 | 9 - 10 | 1648 | 74 | ||
More Information
Set Membership: |
|

PPP2R5A
Genomic Map