Assay Details
| Gene Symbol: FAM3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722647.3 | XP_002722693.1 | 552 | 61 | ||
| XM_002722648.3 | XP_002722694.1 | 439 | 61 | |||
| XM_008250290.2 | XP_008248512.1 | 632 | 61 | |||
| XM_008250291.2 | XP_008248513.1 | 518 | 61 | |||
| XM_017338576.1 | XP_017194065.1 | 636 | 61 | |||
More Information
Set Membership: |
|

FAM3A
Genomic Map