Assay Details
| Gene Symbol: IDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720538.3 | XP_002720584.1 | 1039 | 58 | ||
| XM_008273462.2 | XP_008271684.1 | 1039 | 58 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: IDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720538.3 | XP_002720584.1 | 1039 | 58 | ||
| XM_008273462.2 | XP_008271684.1 | 1039 | 58 | |||
Set Membership: |
|