Assay Details
| Gene Symbol: INAFM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017348372.1 | XP_017203861.1 | 2 | 1068 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

INAFM2| Gene Symbol: INAFM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017348372.1 | XP_017203861.1 | 2 | 1068 | 75 | |
Set Membership: |
|