Assay Details
| Gene Symbol: LOC100352472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008252122.1 | XP_008250344.1 | 2804 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100352472| Gene Symbol: LOC100352472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008252122.1 | XP_008250344.1 | 2804 | 69 | ||
Set Membership: |
|