Assay Details
| Gene Symbol: TMPRSS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249423.2 | XP_008247645.1 | 1066 | 57 | ||
| XM_017338131.1 | XP_017193620.1 | 1071 | 57 | |||
More Information
Set Membership: |
|

TMPRSS4
Genomic Map