Assay Details
| Gene Symbol: CASP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008250692.2 | XP_008248914.1 | 641 | 68 | ||
| XM_008250693.2 | XP_008248915.1 | 591 | 68 | |||
| XM_017338752.1 | XP_017194241.1 | 642 | 68 | |||
| XM_017338753.1 | XP_017194242.1 | 560 | 68 | |||
More Information
Set Membership: |
|

CASP14
Genomic Map