Assay Details
| Gene Symbol: C13H1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715459.3 | XP_002715505.1 | 3 - 4 | 531 | 86 | |
| XM_017345854.1 | XP_017201343.1 | 3 - 4 | 440 | 86 | ||
More Information
Set Membership: |
|

C13H1orf43
Genomic Map