Assay Details
| Gene Symbol: C13H1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715459.3 | XP_002715505.1 | 6 - 7 | 813 | 84 | |
| XM_017345854.1 | XP_017201343.1 | 6 - 7 | 722 | 84 | ||
More Information
Set Membership: |
|

C13H1orf43
Genomic Map