Assay Details
| Gene Symbol: LHX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720474.3 | XP_002720520.1 | 954 | 56 | ||
| XM_008273337.2 | XP_008271559.1 | 978 | 56 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LHX2| Gene Symbol: LHX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720474.3 | XP_002720520.1 | 954 | 56 | ||
| XM_008273337.2 | XP_008271559.1 | 978 | 56 | |||
Set Membership: |
|