Assay Details
| Gene Symbol: BPNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717505.3 | XP_002717551.1 | 8 - 9 | 923 | 66 | |
| XM_008268412.2 | XP_008266634.1 | 9 - 10 | 970 | 66 | ||
| XM_008268413.2 | XP_008266635.1 | 8 - 9 | 833 | 66 | ||
More Information
Set Membership: |
|

BPNT1
Genomic Map