Assay Details
| Gene Symbol: FOXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719607.3 | XP_002719653.1 | 3 - 4 | 918 | 59 | |
| XM_008271934.2 | XP_008270156.1 | 5 - 6 | 1071 | 59 | ||
| XM_008271935.2 | XP_008270157.1 | 6 - 7 | 1147 | 59 | ||
| XM_008271936.2 | XP_008270158.1 | 4 - 5 | 941 | 59 | ||
| XM_008271938.2 | XP_008270160.1 | 5 - 6 | 1071 | 59 | ||
| XM_008271939.2 | XP_008270161.1 | 5 - 6 | 1071 | 59 | ||
| XM_017349385.1 | XP_017204874.1 | 5 - 6 | 940 | 59 | ||
More Information
Set Membership: |
|

FOXN3
Genomic Map