Assay Details
| Gene Symbol: GSDMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719347.3 | XP_002719393.1 | 5 - 6 | 781 | 55 | |
| XM_008271426.2 | XP_008269648.1 | 5 - 6 | 782 | 55 | ||
| XM_017349190.1 | XP_017204679.1 | 5 - 6 | 782 | 55 | ||
More Information
Set Membership: |
|

GSDMA
Genomic Map