Assay Details
| Gene Symbol: SFN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008265896.2 | XP_008264118.1 | 1 | 101 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008265896.2 transcripts.
Set Membership: |
|

SFN
Genomic Map