Assay Details
| Gene Symbol: KRTDAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249521.2 | XP_008247743.1 | 142 | 78 | ||
| XM_008249522.2 | XP_008247744.1 | 143 | 78 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTDAP| Gene Symbol: KRTDAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249521.2 | XP_008247743.1 | 142 | 78 | ||
| XM_008249522.2 | XP_008247744.1 | 143 | 78 | |||
Set Membership: |
|