Assay Details
| Gene Symbol: SRI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713830.3 | XP_002713876.1 | 7 - 8 | 622 | 61 | |
| XM_008261792.2 | XP_008260014.1 | 7 - 8 | 589 | 61 | ||
More Information
Set Membership: |
|

SRI
Genomic Map