Assay Details
| Gene Symbol: RAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017339408.1 | XP_017194897.1 | 369 | 73 | ||
| XM_017339409.1 | XP_017194898.1 | 312 | 73 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAC1| Gene Symbol: RAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017339408.1 | XP_017194897.1 | 369 | 73 | ||
| XM_017339409.1 | XP_017194898.1 | 312 | 73 | |||
Set Membership: |
|