Assay Details
| Gene Symbol: ACER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708078.3 | XP_002708124.1 | 5 - 6 | 795 | 61 | |
| XM_008254272.2 | XP_008252494.1 | 6 - 7 | 724 | 61 | ||
| XM_008254279.2 | XP_008252501.1 | 4 - 5 | 683 | 61 | ||
| XM_017340993.1 | XP_017196482.1 | 5 - 6 | 699 | 61 | ||
| XM_017341001.1 | XP_017196490.1 | 4 - 5 | 561 | 61 | ||
| XM_017341005.1 | XP_017196494.1 | 4 - 5 | 667 | 61 | ||
More Information
Set Membership: |
|

ACER2
Genomic Map