Assay Details
| Gene Symbol: ARF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269632.2 | XP_008267854.1 | 1 - 2 | 273 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008269632.2 transcripts.
Set Membership: |
|

ARF6
Genomic Map