Assay Details
| Gene Symbol: TPM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722089.3 | XP_002722135.1 | 1 | 531 | 54 | |
| XM_008249321.2 | XP_008247543.1 | 539 | 54 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPM4| Gene Symbol: TPM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722089.3 | XP_002722135.1 | 1 | 531 | 54 | |
| XM_008249321.2 | XP_008247543.1 | 539 | 54 | |||
Set Membership: |
|