Assay Details
| Gene Symbol: COL6A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716174.3 | XP_002716220.2 | 33 - 34 | 5960 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

COL6A6| Gene Symbol: COL6A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716174.3 | XP_002716220.2 | 33 - 34 | 5960 | 87 | |
Set Membership: |
|