Assay Details
Gene Symbol: TOGARAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718163.3 | XP_002718209.1 | 10 - 11 | 3889 | 108 | |
XM_008269541.2 | XP_008267763.1 | 11 - 12 | 4055 | 108 | ||
XM_008269542.2 | XP_008267764.1 | 12 - 13 | 4002 | 108 | ||
XM_008269543.2 | XP_008267765.1 | 11 - 12 | 4043 | 108 | ||
XM_008269544.2 | XP_008267766.1 | 10 - 11 | 3902 | 108 | ||
XM_008269546.2 | XP_008267768.1 | 11 - 12 | 3139 | 108 | ||
XM_008269547.2 | XP_008267769.1 | 11 - 12 | 4056 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
