Assay Details
| Gene Symbol: TOGARAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269541.2 | XP_008267763.1 | 8 - 9 | 3631 | 99 | |
| XM_008269542.2 | XP_008267764.1 | 9 - 10 | 3578 | 99 | ||
| XM_008269543.2 | XP_008267765.1 | 8 - 9 | 3619 | 99 | ||
| XM_008269546.2 | XP_008267768.1 | 8 - 9 | 2715 | 99 | ||
| XM_008269547.2 | XP_008267769.1 | 8 - 9 | 3632 | 99 | ||
More Information
Set Membership: |
|

TOGARAM1
Genomic Map