Assay Details
| Gene Symbol: GLYCTK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017343860.1 | XP_017199349.1 | 1 - 2 | 653 | 81 | |
| XM_017343861.1 | XP_017199350.1 | 2 - 3 | 468 | 81 | ||
| XM_017343862.1 | XP_017199351.1 | 3 - 4 | 537 | 81 | ||
| XM_017343863.1 | XP_017199352.1 | 2 - 3 | 465 | 81 | ||
More Information
Set Membership: |
|

GLYCTK
Genomic Map