Assay Details
| Gene Symbol: GK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272484.2 | XP_008270706.1 | 17 - 18 | 1476 | 65 | |
| XM_008272485.2 | XP_008270707.1 | 16 - 17 | 1457 | 65 | ||
| XM_008272486.2 | XP_008270708.1 | 16 - 17 | 1390 | 65 | ||
| XM_008272488.2 | XP_008270710.1 | 15 - 16 | 1371 | 65 | ||
| XM_017349868.1 | XP_017205357.1 | 17 - 18 | 1393 | 65 | ||
| XM_017349869.1 | XP_017205358.1 | 16 - 17 | 1373 | 65 | ||
| XM_017349870.1 | XP_017205359.1 | 17 - 18 | 1392 | 65 | ||
| XM_017349871.1 | XP_017205360.1 | 16 - 17 | 1305 | 65 | ||
| XM_017349872.1 | XP_017205361.1 | 17 - 18 | 1389 | 65 | ||
| XM_017349873.1 | XP_017205362.1 | 15 - 16 | 1285 | 65 | ||
More Information
Set Membership: |
|

GK
Genomic Map