Assay Details
Gene Symbol: C13H1orf162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264672.2 | XP_008262894.1 | 1 - 2 | 302 | 61 | |
XM_008264676.2 | XP_008262898.1 | 1 - 2 | 303 | 61 | ||
XM_008264678.2 | XP_008262900.1 | 1 - 2 | 304 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008264672.2, XM_008264676.2, XM_008264678.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
