Assay Details
| Gene Symbol: LOC100358819 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017349976.1 | XP_017205465.1 | 11 - 12 | 3059 | 98 | |
| XM_017349978.1 | XP_017205467.1 | 10 - 11 | 2888 | 98 | ||
More Information
Set Membership: |
|

LOC100358819
Genomic Map