Assay Details
| Gene Symbol: LOC100358819 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017349976.1 | XP_017205465.1 | 8 - 9 | 2552 | 71 | |
| XM_017349977.1 | XP_017205466.1 | 8 - 9 | 2552 | 71 | ||
| XM_017349978.1 | XP_017205467.1 | 7 - 8 | 2381 | 71 | ||
More Information
Set Membership: |
|

LOC100358819
Genomic Map