Assay Details
| Gene Symbol: PSMB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714610.3 | XP_002714656.2 | 5 - 6 | 1077 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PSMB8| Gene Symbol: PSMB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714610.3 | XP_002714656.2 | 5 - 6 | 1077 | 69 | |
Set Membership: |
|