Assay Details
| Gene Symbol: BTF3L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715097.3 | XP_002715143.1 | 4 - 5 | 522 | 78 | |
| XM_008265254.2 | XP_008263476.1 | 3 - 4 | 545 | 78 | ||
More Information
Set Membership: |
|

BTF3L4
Genomic Map