Assay Details
| Gene Symbol: SPTLC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710970.3 | XP_002711016.1 | 8 - 9 | 1537 | 70 | |
| XM_017341811.1 | XP_017197300.1 | 8 - 9 | 1548 | 70 | ||
More Information
Set Membership: |
|

SPTLC3
Genomic Map