Assay Details
| Gene Symbol: AQP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708029.3 | XP_002708075.1 | 3 - 4 | 432 | 76 | |
| XM_017340827.1 | XP_017196316.1 | 3 - 4 | 437 | 76 | ||
More Information
Set Membership: |
|

AQP3
Genomic Map