Assay Details
| Gene Symbol: TP53I13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718907.3 | XP_002718953.1 | 4 - 5 | 357 | 89 | |
| XM_017348936.1 | XP_017204425.1 | 4 - 5 | 894 | 89 | ||
More Information
Set Membership: |
|

TP53I13
Genomic Map