Assay Details
| Gene Symbol: FBXO31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017339671.1 | XP_017195160.1 | 3352 | 103 | ||
| XM_017339672.1 | XP_017195161.1 | 3265 | 103 | |||
More Information
Set Membership: |
|

FBXO31
Genomic Map